Learn More
Aviva Systems Biology Corp SLC2A5 Antibody (ARP42096_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42096_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC2A5. Alternate names: GLUT5, GLUT-5ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-58105