Learn More
Aviva Systems Biology Corp SLC2A6 Antibody - C-terminal region (ARP43974_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43974_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SLC2A6. Alternate names: GLUT6, GLUT9, HSA011372ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53389