Learn More
Aviva Systems Biology Corp SLC2A9 Antibody - middle region (ARP43757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC2A9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC2A9. Alternate names: GLUT9, GLUTX, UAQTL2, URATv1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58620