Learn More
Aviva Systems Biology Corp SLC30A3 Antibody - middle region (ARP43848_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC30A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC30A3. Alternate names: ZNT3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-62994