Learn More
Aviva Systems Biology Corp SLC30A9 Antibody - N-terminal region (ARP32764_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32764_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC30A9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC30A9. Alternate names: HUEL, ZNT9, GAC63, C4orf1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-54749