Learn More
Aviva Systems Biology Corp SLC34A3 Antibody - middle region (ARP31567_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31567_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC34A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC34A3. Alternate names: FLJ38680, HHRH, NPTIIcApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54269