Learn More
Aviva Systems Biology Corp SLC35A3 Antibody - middle region (ARP43928_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43928_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC35A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC35A3. Alternate names: DKFZp781P1297ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-014-58668