Learn More
Aviva Systems Biology Corp SLC35A4 Antibody - middle region (ARP44089_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44089_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC35A4. It was validated on Western Blot. SLC35A4,solute carrier family 35, member A4,MGC2541; SLC35A4,probable UDP-sugar transporter protein SLC35A4,probable UDP-sugar transporter protein SLC35A4,OTTHUMP00000223644,tumor rejection antigen,solute carrier family 35 (UDP-galactose transporter), member A4The immunogen is a synthetic peptide directed towards the middle region of human SLC35A4. Alternate names: MGC2541ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Rabbit, Yeast
Catalog No. 50-285-123