Learn More
Aviva Systems Biology Corp SLC35A5 Antibody - N-terminal region (ARP43979_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43979_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC35A5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC35A5. Alternate names: DKFZp434E102, FLJ11130, FLJ20730, FLJ25973ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58684