Learn More
Aviva Systems Biology Corp SLC35C1 Antibody - N-terminal region (ARP43989_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43989_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC35C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC35C1. Alternate names: FLJ11320, FLJ14841, FUCT1, CDG2CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53390