Learn More
Aviva Systems Biology Corp SLC35F2 Antibody - N-terminal region (ARP43971_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43971_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC35F2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC35F2. Alternate names: DKFZp667H1615, FLJ13018, HSNOV1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-58679