Learn More
Aviva Systems Biology Corp Slc35f3 Antibody - C-terminal region (ARP44141_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44141_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc35f3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: B230375D17Rik, MGC141263, MGC141264ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-124