Learn More
Aviva Systems Biology Corp SLC35F5 Antibody - C-terminal region (ARP44044_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44044_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC35F5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SLC35F5. Alternate names: FLJ22004ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53395