Learn More
Aviva Systems Biology Corp SLC36A1 Antibody - N-terminal region (ARP64286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC36A1. It was validated on Western Blot. . Alternate names: Dct1, LYAAT1, PAT1, TRAMD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-222