Learn More
Aviva Systems Biology Corp SLC37A3 Antibody - middle region (ARP44064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC37A3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC37A3. Alternate names: MGC32939ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-58705