Learn More
Aviva Systems Biology Corp SLC37A4 Antibody - middle region (ARP43821_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43821_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC37A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC37A4. Alternate names: G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, MGC15729, PRO0685, TRG19, TRG-19ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58635