Learn More
Aviva Systems Biology Corp SLC38A1 Antibody - middle region (ARP43810_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43810_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC38A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC38A1. Alternate names: ATA1, NAT2, SAT1, SNAT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-53377