Learn More
Aviva Systems Biology Corp SLC38A3 Antibody - N-terminal region (ARP42323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC38A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC38A3. Alternate names: G17, SN1, NAT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58183