Learn More
Aviva Systems Biology Corp SLC38A4 Antibody - middle region (ARP42504_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42504_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC38A4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC38A4. Alternate names: ATA3, FLJ10191, MGC126876, NAT3, PAATApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53341