Learn More
Aviva Systems Biology Corp SLC38A5 Antibody - N-terminal region (ARP44075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC38A5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC38A5. Alternate names: JM24, SN2, pp7194, SNAT5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58708