Learn More
Aviva Systems Biology Corp SLC39A12 Antibody - N-terminal region (ARP44127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC39A12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC39A12. Alternate names: FLJ30499, MGC43205, MGC51099, bA570F3.1, ZIP-12, LZT-Hs8ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-58726