Learn More
Aviva Systems Biology Corp SLC39A4 Antibody - N-terminal region (ARP44090_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44090_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC39A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC39A4. Alternate names: AEZ, FLJ20327, MGC74741, ZIP4, AWMS2ApplicationWB. Species reactivity: Cow, Human, Rabbit
Catalog No. 50-014-58714