Learn More
Biomatik Corporation Slc39A4 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10802
Encompass_Preferred
Slc39A4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC39A4. The Slc39A4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Transporter Zip4 Protein (23-327Aa). Applications: ELISA, WB, IHC. Target Synonyms: SLC39A4. Accession Number: Q6P5W5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4978