Learn More
Aviva Systems Biology Corp SLC39A5 Antibody - N-terminal region (ARP44143_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44143_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC39A5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC39A5. Alternate names: LZT-Hs7, MGC34778, ZIP5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53407