Learn More
Aviva Systems Biology Corp SLC39A6 Antibody - middle region (ARP43931_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43931_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC39A6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC39A6. Alternate names: LIV-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53385