Learn More
Aviva Systems Biology Corp SLC39A8 Antibody - N-terminal region (ARP44032_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44032_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC39A8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC39A8. Alternate names: BIGM103, LZT-Hs6, ZIP8, PP3105ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-58696