Learn More
Aviva Systems Biology Corp SLC3A1 Antibody - N-terminal region (ARP43743_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43743_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC3A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC3A1. Alternate names: ATR1, CSNU1, D2H, FLJ34681, NBAT, RBATApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58615