Learn More
Sino Biological SLC3A2 / CD98 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 50813R048100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SLC3A2 (rM SLC3A2; Catalog#50813-M07H; NP_001154885.1; Ala139-Ala565).
Catalog No. 50-222-3355