Learn More
Aviva Systems Biology Corp SLC40A1 Antibody - middle region (ARP43953_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43953_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC40A1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SLC40A1. Alternate names: FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-113