Learn More
Aviva Systems Biology Corp SLC41A2 Antibody - N-terminal region (ARP44062_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44062_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC41A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC41A2. Alternate names: DKFZP434K0427, MGC125330, MGC125331, SLC41A1-L1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-53399