Learn More
Aviva Systems Biology Corp SLC43A1 Antibody - middle region (ARP43854_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43854_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC43A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC43A1. Alternate names: LAT3, PB39, POV1, R00504ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-58648