Learn More
Aviva Systems Biology Corp SLC43A2 Antibody - N-terminal region (ARP44116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC43A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC43A2. Alternate names: FLJ23848, LAT4, MGC34680ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-53404