Learn More
Aviva Systems Biology Corp SLC43A3 Antibody - N-terminal region (ARP43938_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43938_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC43A3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC43A3. Alternate names: DKFZp762A227, EEG1, FOAP-13, PRO1659, SEEEG-1ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-58671