Learn More
Aviva Systems Biology Corp SLC45A1 Antibody - N-terminal region (ARP44193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC45A1. It was validated on Western Blot. . Alternate names: DNB5, FLJ26758, KIAA0458ApplicationWB. Species reactivity: Human
Catalog No. 50-285-128