Learn More
Aviva Systems Biology Corp SLC45A3 Antibody - middle region (ARP44071_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44071_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC45A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC45A3. Alternate names: IPCA-6, IPCA6, PCANAP6, PRST, IPCA-2, IPCA-8, PCANAP2, PCANAP8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53400