Learn More
Aviva Systems Biology Corp SLC46A3 Antibody - N-terminal region (ARP44158_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44158_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC46A3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC46A3. Alternate names: FKSG16, RP11-97E23.2ApplicationIHC, WB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58735