Learn More
Aviva Systems Biology Corp SLC4A1 Antibody - N-terminal region (ARP33801_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC4A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLC4A1. Alternate names: DI, FR, SW, WD, WR, AE1, CHC, SAO, WD1, BND3, EPB3, SPH4, CD233, EMPB3, RTA1AApplicationWB. Species reactivity: Horse, Human, Rabbit
Catalog No. 50-014-55151