Learn More
Aviva Systems Biology Corp SLC4A5 Antibody - middle region (ARP44021_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44021_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC4A5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC4A5. Alternate names: MGC129662, NBC4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58693