Learn More
ABclonal Technology SLC4A8 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148251000UL
Encompass_Preferred
The protein encoded by this gene is a membrane protein that functions to transport sodium and bicarbonate ions across the cell membrane. The encoded protein is important for pH regulation in neurons. The activity of this protein can be inhibited by 4,4-Di-isothiocyanatostilbene-2,2-disulfonic acid (DIDS). Several transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07125