Learn More
Aviva Systems Biology Corp Slc5a2 Antibody - middle region (ARP43832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc5a2. It was validated on Western Blot. . Alternate names: Sglt2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-102