Learn More
Aviva Systems Biology Corp SLC5A7 Antibody - middle region (ARP44024_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44024_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC5A7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC5A7. Alternate names: CHT, CHT1, MGC126299, MGC126300, hCHTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58694