Learn More
Aviva Systems Biology Corp SLC6A1 Antibody - middle region (ARP43834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC6A1. Alternate names: GABATHG, GABATR, GAT1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58640