Learn More
Aviva Systems Biology Corp SLC6A14 Antibody - middle region (ARP43925_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43925_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC6A14. Alternate names: ATB(0+), BMIQ11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58667