Learn More
Aviva Systems Biology Corp Slc6a15 Antibody - middle region (ARP44162_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc6a15. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA536730, AI326450, AI326451, v7-3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-125