Learn More
Aviva Systems Biology Corp SLC6A15 Antibody - middle region (ARP44163_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44163_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC6A15. Alternate names: DKFZp761I0921, FLJ10316, MGC87066, NTT73, V7-3, hv7-3, SBAT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62997