Learn More
Aviva Systems Biology Corp SLC6A18 Antibody - middle region (ARP44161_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44161_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A18. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SLC6A18. Alternate names: FLJ31236, Xtrp2ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58736