Learn More
Aviva Systems Biology Corp SLC6A2 Antibody - middle region (ARP41893_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41893_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC6A2. Alternate names: NAT1, NET, NET1, SLC6A5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58036