Learn More
Aviva Systems Biology Corp SLC6A3 Antibody - N-terminal region (ARP43791_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43791_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SLC6A3. Alternate names: DAT, DAT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-097