Learn More
Aviva Systems Biology Corp SLC6A8 Antibody - middle region (ARP42249_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLC6A8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLC6A8. Alternate names: CRTR, CT1, MGC87396, CRTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58162