Learn More
Aviva Systems Biology Corp Slc6a9 Antibody - C-terminal region (ARP42330_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42330_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Slc6a9. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Glyt-1, Glyt1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-993